KEY371462915 Key Organics / BIONET

2,6-diamino-5-bromopyrimidin-4-ol

  • MDL number MFCD00023262
  • Cas Number : 6312-72-7
  • Molecular Weight : 205.015

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 256 - 257
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97570

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GG-0242_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart