KEY371462927 Key Organics / BIONET

3-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]amino}propane-1,2-diol

  • MDL number MFCD22383956
  • Cas Number : 1393845-79-8
  • Molecular Weight : 270.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102302

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MF-0049_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart