KEY371462931 Key Organics / BIONET

1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD00020785
  • Cas Number : 83-10-3
  • Molecular Weight : 232.239

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108379

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3597_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart