KEY371462932 Key Organics / BIONET

tert-butyl N-(2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-3-yl)carbamate

  • MDL number MFCD19441845
  • Cas Number : 86499-69-6
  • Molecular Weight : 276.336

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108389

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3682_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart