KEY371462939 Key Organics / BIONET

ethyl 2-(4,6-dihydroxypyrimidin-5-yl)acetate

  • MDL number MFCD20527459
  • Cas Number : 1095822-20-0
  • Molecular Weight : 198.178

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108623

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4344_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart