KEY371462942 Key Organics / BIONET

6-[(tert-butyldimethylsilyl)oxy]-1H-indole

  • MDL number MFCD02683922
  • Cas Number : 106792-41-0
  • Molecular Weight : 247.413

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108626

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4348_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart