KEY371462947 Key Organics / BIONET

(2E)-2-{[4-methoxy-3-(3-methoxypropoxy)phenyl]methylidene}-3-methylbutanoic acid

  • MDL number MFCD11112069
  • Cas Number : 387868-07-7
  • Molecular Weight : 308.374

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108667

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4390_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart