KEY375226979 Key Organics / BIONET

methyl 1-benzofuran-2-carboxylate

  • MDL number MFCD01159618
  • Cas Number : 1646-27-1
  • Molecular Weight : 176.171

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53 - 55
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90475

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8R-0046_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart