KEY375226980 Key Organics / BIONET

N-(4-fluoro-2-methyl-6-nitrophenyl)acetamide

  • MDL number MFCD11520129
  • Cas Number : 159020-73-2
  • Molecular Weight : 212.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170 - 172
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94473

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AB-0824_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart