KEY375226981 Key Organics / BIONET

5,6-diamino-1,3-diethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione hydrochloride

  • MDL number MFCD22573561
  • Cas Number : 1785764-26-2
  • Molecular Weight : 234.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 270
Iupac Name 5,6-diamino-1,3-diethyl-2,4(1H,3H)-pyrimidinedione hydrochloride
Inchi Code 1S/C8H14N4O2.ClH/c1-3-11-6(10)5(9)7(13)12(4-2)8(11)14;/h3-4,9-10H2,1-2H3;1H
Inchi Key UQWLJOFXKBZWCI-UHFFFAOYSA-N

Description

Melting Point 270
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94842

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-0045_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart