KEY375226984 Key Organics / BIONET

2,4-dimethyl 3-amino-1H-pyrrole-2,4-dicarboxylate

  • MDL number MFCD10566520
  • Cas Number : 180059-04-5
  • Molecular Weight : 198.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 165 - 166
Iupac Name dimethyl 3-amino-1H-pyrrole-2,4-dicarboxylate
Inchi Code 1S/C8H10N2O4/c1-13-7(11)4-3-10-6(5(4)9)8(12)14-2/h3,10H,9H2,1-2H3
Inchi Key VFKYODDQOZLTQL-UHFFFAOYSA-N

Description

Melting Point 165 - 166
Purity 97%
Storage temp. Store under N2
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97150

Safety Information

signalword Warning
pcodes P422, P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/FG-0711_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart