KEY375226986 Key Organics / BIONET

methyl 4-({8-chloro-2-methyl-1H,2H,3H,4H,5H-pyrido[4,3-b]indol-5-yl}methyl)benzoate

  • MDL number MFCD22573514
  • Cas Number : 1354547-26-4
  • Molecular Weight : 368.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132 - 133
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97568

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GG-0209_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart