KEY375226990 Key Organics / BIONET

1-bromo-2-(chloromethyl)-4-fluorobenzene

  • MDL number MFCD09263441
  • Cas Number : 857276-61-0
  • Molecular Weight : 223.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97957

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/HG-0023_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart