KEY375226991 Key Organics / BIONET

methyl (2E)-2-(N-hydroxyimino)-3-(4-hydroxyphenyl)propanoate

  • MDL number MFCD22573515
  • Cas Number : 50563-23-0
  • Molecular Weight : 209.201

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131 - 132
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97959

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/HG-0044_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart