KEY375226994 Key Organics / BIONET

tert-butyl 5-benzyl-octahydropyrrolo[3,4-c]pyrrole-2-carboxylate

  • MDL number MFCD18909636
  • Cas Number : 186202-73-3
  • Molecular Weight : 302.418

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil not distilled
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97962

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/HG-0052_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart