KEY375227004 Key Organics / BIONET

tert-butyl octahydropyrrolo[3,4-c]pyrrole-2-carboxylate

  • MDL number MFCD04115128
  • Cas Number : 141449-85-6
  • Molecular Weight : 212.293

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110 - 112
Purity 97
Storage temp. RT
Physical Form White to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101117

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/KG-0017_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart