KEY375227018 Key Organics / BIONET

methyl 5-bromo-4-hydroxy-[1,1'-biphenyl]-3-carboxylate

  • MDL number MFCD22573529
  • Cas Number : 1155261-80-5
  • Molecular Weight : 307.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102017

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/LG-0018_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart