KEY375227042 Key Organics / BIONET

methyl 7-(trifluoromethyl)-1-benzofuran-2-carboxylate

  • MDL number MFCD22573541
  • Cas Number : 1407521-92-9
  • Molecular Weight : 244.169

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51 - 53
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102047

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/LG-0745_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart