KEY375227046 Key Organics / BIONET

1-(4-fluorophenyl)-4-oxocyclohexane-1-carbonitrile

  • MDL number MFCD00044815
  • Cas Number : 56326-98-8
  • Molecular Weight : 217.243

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108377

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3591_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart