KEY375227048 Key Organics / BIONET

2,3-dihydro-1-benzofuran-7-carbaldehyde

  • MDL number MFCD06200853
  • Cas Number : 196799-45-8
  • Molecular Weight : 148.161

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62 - 64
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108390

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3697_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart