KEY375227052 Key Organics / BIONET

2-[(1,1'-biphenyl)-3-yl]ethanol

  • MDL number MFCD09926177
  • Cas Number : 71912-71-5
  • Molecular Weight : 198.265

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108641

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4364_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart