KEY375227054 Key Organics / BIONET

4-bromo-N-(1,3-thiazol-2-yl)benzene-1-sulfonamide

  • MDL number MFCD00592646
  • Cas Number : 331972-47-5
  • Molecular Weight : 319.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108675

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4400_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart