KEY375227056 Key Organics / BIONET

8-bromo-2-methoxy-1,5-naphthyridine

  • MDL number MFCD09027616
  • Cas Number : 881658-92-0
  • Molecular Weight : 239.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-bromo-2-methoxy[1,5]naphthyridine
Inchi Code 1S/C9H7BrN2O/c1-13-8-3-2-7-9(12-8)6(10)4-5-11-7/h2-5H,1H3
Inchi Key HPQMRRPAOIGXFW-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108677

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4403_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart