KEY375227058 Key Organics / BIONET

methyl (3E)-4-(2-bromophenyl)-2-oxobut-3-enoate

  • MDL number MFCD22573543
  • Cas Number : 956476-32-7
  • Molecular Weight : 269.094

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108679

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4407_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart