KEY375227069 Key Organics / BIONET

1-[(4-fluorophenyl)methyl]-1H-indazole-3-carboxylic acid

  • MDL number MFCD01719717
  • Cas Number : 50264-63-6
  • Molecular Weight : 270.263

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108692

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4426_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart