KEY375227073 Key Organics / BIONET

tert-butyl 3-iodo-1H-indazole-1-carboxylate

  • MDL number MFCD09835520
  • Cas Number : 290368-00-2
  • Molecular Weight : 344.152

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108697

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4437_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart