KEY375227075 Key Organics / BIONET

4-bromo-N-(1,3,4-thiadiazol-2-yl)benzene-1-sulfonamide

  • MDL number MFCD02861948
  • Cas Number : 349614-28-4
  • Molecular Weight : 320.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108699

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4440_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart