KEY375227078 Key Organics / BIONET

methyl 4-(2H-1,2,3,4-tetrazol-5-yl)benzoate

  • MDL number MFCD20441656
  • Cas Number : 82544-82-9
  • Molecular Weight : 204.189

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108702

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4444_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart