KEY375227084 Key Organics / BIONET

tert-butyl 11-azatricyclo[6.2.1.0^{2,7}]undeca- 2,4,6,9-tetraene-11-carboxylate

  • MDL number MFCD22573547
  • Cas Number : 5176-28-3
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 1,4-dihydro-1,4-epiminonaphthalene-9-carboxylate
Inchi Code 1S/C15H17NO2/c1-15(2,3)18-14(17)16-12-8-9-13(16)11-7-5-4-6-10(11)12/h4-9,12-13H,1-3H3
Inchi Key KTZZRAGNIAZODS-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108709

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4455_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart