KEY375227086 Key Organics / BIONET

5-bromo-[1,3]thiazolo[5,4-b]pyridin-2-amine

  • MDL number MFCD11846492
  • Cas Number : 934266-82-7
  • Molecular Weight : 230.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108711

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4460_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart