KEY375227087 Key Organics / BIONET

methyl 2-(4-hydroxy-2-methoxyphenyl)acetate

  • MDL number MFCD18399380
  • Cas Number : 499789-92-3
  • Molecular Weight : 196.202

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108714

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4464_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart