KEY387783358 Key Organics / BIONET

2,3,4,5,6-pentafluorophenyl [1,1'-biphenyl]-4-sulfonate

  • MDL number MFCD22689145
  • Cas Number : 1420537-69-4
  • Molecular Weight : 400.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7280

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11X-0266_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart