KEY387783361 Key Organics / BIONET

2-(4-nitrophenyl)-4H,5H,6H,7H-furo[3,2-c]pyridine

  • MDL number MFCD22689166
  • Cas Number : 1192968-13-0
  • Molecular Weight : 244.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108365

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3482_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart