KEY387783362 Key Organics / BIONET

(1R)-3-(dimethylamino)-1-phenylpropan-1-ol; oxalic acid

  • MDL number MFCD22689168
  • Cas Number : 1270016-44-8
  • Molecular Weight : 269.297

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108776

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4527_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart