KEY387783367 Key Organics / BIONET

1-bromo-3,5-bis(bromomethyl)benzene

  • MDL number MFCD00192715
  • Cas Number : 51760-23-7
  • Molecular Weight : 342.856

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95845

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CG-0013_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart