KEY387783383 Key Organics / BIONET

5-(2-methoxyphenyl)-4-(4-methoxyphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD02246710
  • Cas Number : 483283-17-6
  • Molecular Weight : 313.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 244 - 245
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102340

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MG-0203_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart