KEY387783394 Key Organics / BIONET

(3R)-3-{[(tert-butoxy)carbonyl]amino}-4-(2,4,5-trifluorophenyl)butanoic acid

  • MDL number MFCD06659147
  • Cas Number : 486460-00-8
  • Molecular Weight : 333.307

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102355

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/MG-0718_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart