KEY387783418 Key Organics / BIONET

(2S)-2-amino-3-methylbutan-1-ol hydrochloride

  • MDL number MFCD00050559
  • Cas Number : 17016-89-6
  • Molecular Weight : 139.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2S)-2-amino-3-methyl-1-butanol hydrochloride
Inchi Code 1S/C5H13NO.ClH/c1-4(2)5(6)3-7;/h4-5,7H,3,6H2,1-2H3;1H/t5-;/m1./s1
Inchi Key CYONGLLQMUYOPP-NUBCRITNSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108572

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4171_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart