KEY387783419 Key Organics / BIONET

9-(pyrrolidin-1-ylmethyl)-9H-carbazole

  • MDL number MFCD00958842
  • Cas Number : 111960-23-7
  • Molecular Weight : 250.345

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74 - 75
Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108633

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4356_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart