KEY387783426 Key Organics / BIONET

(2S)-3-amino-2-methylpropan-1-ol

  • MDL number MFCD19203687
  • Cas Number : 88586-62-3
  • Molecular Weight : 89.138

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108671

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4394_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart