KEY387783444 Key Organics / BIONET

5-amino-3-methyl-1-phenyl-1H-pyrazole-4-carbonitrile

  • MDL number MFCD00128285
  • Cas Number : 5346-56-5
  • Molecular Weight : 198.229

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108783

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4534_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart