KEY387783445 Key Organics / BIONET

2-bromo-5-(pyrrolidine-1-carbonyl)-1,3-thiazole

  • MDL number MFCD20259835
  • Cas Number : 877675-06-4
  • Molecular Weight : 261.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108784

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4535_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart