KEY387783447 Key Organics / BIONET

methyl (3E)-4-(4-bromophenyl)-2-oxobut-3-enoate

  • MDL number MFCD18642374
  • Cas Number : 608128-34-3
  • Molecular Weight : 269.094

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108786

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4537_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart