KEY387783449 Key Organics / BIONET

5-bromo-3-chloro-6-(trifluoromethyl)-1H-indole

  • MDL number MFCD22689147
  • Cas Number : 1420537-62-7
  • Molecular Weight : 298.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 88 - 91
Iupac Name 5-bromo-3-chloro-6-(trifluoromethyl)-1H-indole
Inchi Code 1S/C9H4BrClF3N/c10-6-1-4-7(11)3-15-8(4)2-5(6)9(12,13)14/h1-3,15H
Inchi Key SWTSRWZDZNHRAZ-UHFFFAOYSA-N

Description

Melting Point 88 - 91
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94812

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/AH-0701_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart