KEY387783451 Key Organics / BIONET

5-chloro-7-nitro-2,3-dihydro-1,3-benzoxazol-2-one

  • MDL number MFCD09753602
  • Cas Number : 811810-67-0
  • Molecular Weight : 214.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108790

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4541_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart