KEY387783455 Key Organics / BIONET

4-bromo-7-methoxy-2,2-dimethyl-2,3-dihydro-1-benzofuran

  • MDL number MFCD22414513
  • Cas Number : 185406-76-2
  • Molecular Weight : 257.127

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108794

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4546_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart