KEY387783460 Key Organics / BIONET

2-{[(tert-butoxy)carbonyl]amino}-3-(trifluoromethoxy)benzoic acid

  • MDL number MFCD09800581
  • Cas Number : 561304-40-3
  • Molecular Weight : 321.252

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108799

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4551_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart