KEY387783463 Key Organics / BIONET

ethyl 2-(2-amino-2,3-dihydro-1H-inden-2-yl)acetate hydrochloride

  • MDL number MFCD22689175
  • Cas Number : 917391-08-3
  • Molecular Weight : 255.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108802

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4554_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart