KEY387783465 Key Organics / BIONET

8-hydroxy-1H,2H,3H,4H-cyclopenta[c]chromen-4-one

  • MDL number MFCD08691761
  • Cas Number : 131526-88-0
  • Molecular Weight : 202.209

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108804

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4556_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart