KEY387783466 Key Organics / BIONET

2,2-dimethyl-6-nitro-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one

  • MDL number MFCD10000811
  • Cas Number : 1002726-59-1
  • Molecular Weight : 223.188

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108805

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4558_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart